Gold tribromide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Gold (III) bromide, Gold(III) bromide

A19514-06 A19514-03
AAAA19514-06EA 930 USD
AAAA19514-06 AAAA19514-03
Gold tribromide 99%
Gold tribromide

Purity: 99% CAS No.: 10294-28-7 Molecular Formula: AuBr3 Formula Weight: 436.69 Melting Point: ca 160° dec. UN No.: 3260 MDL No.: MFCD00014171

Formula: AuBr₃
MW: 436.68
Melting Pt: 160 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00014171
CAS Number: 10294-28-7
EINECS: 233-654-2
UN: 3260
ADR: 8 III
Merck Index: 12,04539

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR