2-(2-Hydroxyphenyl)benzothiazole 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(1,3-Benzothiazol-2-yl)phenol

A19346-06 A19346-14
AAA19346-06EA 37 USD
AAA19346-06 AAAA19346-22 AAA19346-14
2-(2-Hydroxyphenyl)benzothiazole 98%
2-(2-Hydroxyphenyl)benzothiazole

Purity: 98% CAS No.: 3411-95-8 Molecular Formula: C13H9NOS Formula Weight: 227.29 Melting Point: 130-134° Beilstein Reference: 173026

Formula: C₁₃H₉NOS
MW: 227.29 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00022869
CAS Number: 3411-95-8
EINECS: 222-299-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR