Butyltin hydroxide oxide 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A18290-22
A18290-36
AAAA18290-22EA
71.9
USD
AAAA18290-22
AAAA18290-0E
AAAA18290-36
Butyltin hydroxide oxide 95%
Butyltin hydroxide oxide
Purity: 95%
CAS No.: 2273-43-0
Molecular Formula: CH3(CH2)3Sn(=O)OH
Formula Weight: 208.81
UN No.: 3146
MDL No.: MFCD00013927
Formula:
C₄H₁₀O₂Sn Storage Temperature: Ambient |
MDL Number:
MFCD00013927 CAS Number: 2273-43-0 EINECS: 218-880-1 UN: 3146 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...