Diphenylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Phenylaniline, N-Phenylbenzeneamine

A18265-36 A18265-22 A18265-0E
AAAA18265-36EA 59.56 USD
AAAA18265-36 AAAA18265-22 AAAA18265-0E
Diphenylamine 98+%
Diphenylamine

Purity: 98+% CAS No.: 122-39-4 Molecular Formula: C12H11N Formula Weight: 169.23 Flash Point: 152°(305°F) Odor: Weak Boiling Point: 302° Beilstein Reference: 508755 Merck Reference: 14,3317 UN No.: 3077 MDL No.: MFCD00003014

Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C
Melting Pt: 52…55 °C
Density: 1.165 g/cm³ (20 °C)
Flash Pt: 152 °C (305 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III
Merck Index: 13,03349
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23b05bdc0d251be7093ced24f3f9389bc8e8d3c5&vid=f543cfadbe22d37b18bd37917bb8b5672a6d2df4&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR