Tin(II) oxide 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Stannous oxide, Tin monoxide, Tin(ii)oxide

A17889-0E A17889-22 A17889-36
AAA17889-0EEA 616 USD
AAA17889-0E AAAA17889-22 AAAA17889-36
Tin(II) oxide 99%
Tin(II) oxide

Purity: 99% CAS No.: 21651-19-4 Molecular Formula: OSn Formula Weight: 134.69 Odor: Odorless Melting Point: 1080° dec. MDL No.: MFCD00011243

Formula: OSn
MW: 134.71 g/mol
Boiling Pt: 1425
Melting Pt: 1040 (dec.)
Density: 6.45
Storage Temperature: Ambient
MDL Number: MFCD00011243
CAS Number: 21651-19-4
EINECS: 244-499-5
Merck Index: 14,8787

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR