Gibberellic acid 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
GA3, Gibberellin A3, (3S,3aS,4S,4aS,6S,8aR,8bR,11S)-6,11-Dihydroxy-3-methyl-12-methylene-2-oxo-4a,6-ethano-3,8b-prop-1-enoperhydroindeno[1,2-b]furan-4-carboxylic acid
A17843-03
A17843-14
A17843-06
AAAA17843-03EA
53.49
USD
AAAA17843-03
AAAA17843-14
AAAA17843-06
Gibberellic acid 90+%
Gibberellic acid
Plant growth hormone
Purity: 90+%
CAS No.: 77-06-5
Molecular Formula: C19H22O6
Formula Weight: 346.37
Melting Point: 222° dec.
Beilstein Reference: 54346
Merck Reference: 14,4419
Optical Rotation: +78° (c=2 in methanol)
MDL No.: MFCD00079329
Formula:
C₁₉H₂₂O₆ MW: 346.38 g/mol Melting Pt: 234 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00079329 CAS Number: 77-06-5 EINECS: 201-001-0 Merck Index: 14,4419 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...