Quinhydrone 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzoquinone-hydroquinone, p-Benzoquinone hydroquinone complex, 2,5-Cyclohexadiene-1,4-dione, compd. with 1,4-benzenediol (1:1)

A16681-14 A16681-22
AAA-A16681-14EA 55.24 USD
AAA-A16681-14 AAA-A16681-22
Quinhydrone 97%
Quinhydrone
Formula: C₆H₄(OH)₂·C₆H₄O₂
MW: 218.21 g/mol
Melting Pt: 168…171 °C
Density: 1.35 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010310
CAS Number: 106-34-3
EINECS: 203-387-6
UN: 2811
ADR: 6.1,III
Merck Index: 13,08148

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR