Mercury(II) sulphate 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

A16330-30 A16330-0B A16330-18
AAAA16330-30EA 172 USD
AAAA16330-30 AAAA16330-0B AAAA16330-18
Mercury(II) sulphate 98+%
Mercury(II) sulphate

Purity: 98+% CAS No.: 7783-35-9 Molecular Formula: HgO4S Formula Weight: 296.65 Melting Point: dec. Merck Reference: 14,5887 MDL No.: MFCD00011047

Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850 Dec.
Density: 6.470
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 14,5887

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR