4-Hydroxy-4-methyl-2-pentanone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diacetone alcohol
A16248-AE
A16248-AP
A16248-0F
AAAA16248-AEEA
38.42
USD
AAAA16248-AE
AAAA16248-AP
AAAA16248-0F
4-Hydroxy-4-methyl-2-pentanone 98+%
4-Hydroxy-4-methyl-2-pentanone
Purity: 98+%
CAS No.: 123-42-2
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 58°(136°F)
Refractive Index: 1.4230
Odor: Pleasant
Melting Point: -47°
Boiling Point: 168°
Beilstein Reference: 1740440
Merck Reference: 14,2964
UN No.: 1148
MDL No.: MFCD00004471
Formula:
(CH₃)₂C(OH)CH₂COCH₃ Boiling Pt: 166 °C (1013 hPa) Melting Pt: -44 °C Density: 0.94 g/cm³ (20 °C) Flash Pt: 58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004471 CAS Number: 123-42-2 EINECS: 204-626-7 UN: 1148 ADR: 3,III REACH: 01-2119473975-21 Merck Index: 13,02983 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...