Mercury (II) oxide, red 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16157-18
A16157-0B
A16157-30
AAAA16157-18EA
34.1
USD
AAAA16157-18
AAAA16157-0B
AAAA16157-30
Mercury (II) oxide, red 99%
Mercury (II) oxide
Purity: 99%
CAS No.: 21908-53-2
Molecular Formula: HgO
Formula Weight: 216.59
Odor: Odorless
Melting Point: ca 500° dec.
UN No.: 1641
MDL No.: MFCD00011045
Formula:
HgO MW: 216.59 g/mol Melting Pt: ∼ 500 °C (dec.) Density: 11.14 Storage Temperature: Ambient |
MDL Number:
MFCD00011045 CAS Number: 21908-53-2 EINECS: 244-654-7 UN: 1641 ADR: 6.1 II Merck Index: 14,5882 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...