Benzenesulfonic acid 94%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A16148-36
A16148-22
A16148-0E
AAAA16148-36EA
39
USD
AAAA16148-36
AAA16148-22
AAAA16148-0E
Benzenesulfonic acid 94%
Benzenesulphonic acid
Acid catalyst for direct esterification of amino acids and peptides
Purity: 94%
CAS No.: 98-11-3
Molecular Formula: C6H6O3S
Formula Weight: 158.18
Flash Point:
Formula:
C₆H₆O₃S MW: 158.18 g/mol Boiling Pt: 319 °C (1013 hPa) Melting Pt: ca 45…50 °C Density: 1.32 g/cm³ (47 °C) Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011689 CAS Number: 98-11-3 EINECS: 202-638-7 UN: 2585 ADR: "8",III Merck Index: 13,01070 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...