Lithium metaborate, (dried basis) 99%

Supplier: Thermo Scientific Chemicals

A16145-14 A16145-36 A16145-22
AAA16145-14EA 39.6 USD
AAA16145-14 AAA16145-36 AAA16145-22
Lithium metaborate, (dried basis) 99%
Lithium metaborate

Purity: 99% CAS No.: 13453-69-5 Molecular Formula: BLiO2 Formula Weight: 49.75 Odor: Odorless Melting Point: 845° MDL No.: MFCD00011089

Formula: BLiO₂
Boiling Pt: 1719 °C (1013 hPa)
Melting Pt: 845 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011089
CAS Number: 13453-69-5
EINECS: 236-631-5
Merck Index: 13,05546

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR