Phenazine 99+%

Supplier: Thermo Scientific Chemicals
Warning

A15770-14 A15770-22 A15770-06
AAAA15770-14EA 145 USD
AAAA15770-14 AAAA15770-22 AAAA15770-06
Phenazine 99+%
Phenazine

Purity: 99+% CAS No.: 92-82-0 Molecular Formula: C12H8N2 Formula Weight: 180.21 Melting Point: 175-177° MDL No.: MFCD00005023

Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: 339 °C (1013 hPa)
Melting Pt: 172 - 174 °C
Density: 1.26 g/cm³ (22 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005023
CAS Number: 92-82-0
EINECS: 202-193-9
Merck Index: 12,07359

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR