4-Fluorophenylhydrazinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Fluorphenyl)hydrazin HCl, 4-Fluorophenylhydrazine hydrochloride, 1-(4-Fluorophenyl)hydrazine hydrochloride, (4-Fluorophenyl)hydrazine hydrochloride
A15624-14
A15624-22
A15624-06
AAA15624-14EA
127
USD
AAA15624-14
AAA15624-22
AAA15624-06
4-Fluorophenylhydrazinium chloride 98%
4-Fluorophenylhydrazinium chloride
Purity: 98%
CAS No.: 823-85-8
Molecular Formula: C6H7FN2.HCl
Formula Weight: 162.60
Melting Point: ca 265° dec.
Beilstein Reference: 3627721
Formula:
C₆H₈ClFN₂ Melting Pt: ca 265 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012942 CAS Number: 823-85-8 EINECS: 212-521-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...