4-Bromophenylhydrazinium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

A15590-14 A15590-06
AAA15590-14EA 229 USD
AAA15590-14 AAA15590-06
4-Bromophenylhydrazinium chloride 97%
4-Bromophenylhydrazinium chloride

Purity: 97% CAS No.: 622-88-8 Molecular Formula: C6H7BrN2.HCl Formula Weight: 223.51 Odor: Odorless Melting Point: ca 215° dec. Beilstein Reference: 3565838 MDL No.: MFCD00012941

Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Melting Pt: ∼ 215 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012941
CAS Number: 622-88-8
EINECS: 255-590-4
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR