2-Butynoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15542-06
A15542-14
AAAA15542-06EA
148
USD
AAAA15542-06
AAAA15542-14
2-Butynoic acid 98%
2-Butynoic acid
Purity: 98%
CAS No.: 590-93-2
Molecular Formula: C4H4O2
Formula Weight: 84.07
Storage Temperature: 2°C to 8°C
Melting Point: 75-79°
Boiling Point: 200-203°
Beilstein Reference: 1740205
UN No.: 3261
MDL No.: MFCD00004363
Formula:
C₄H₄O₂ MW: 84.07 g/mol Boiling Pt: 200…203 °C Melting Pt: 75…79 °C Density: 0.9641 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004363 CAS Number: 590-93-2 EINECS: 209-695-7 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...