2-Amino-3-bromo-5-methylbenzoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15443-06
A15443-14
AAA15443-03EA
32.25
USD
AAA15443-03
AAAA15443-06
AAAA15443-14
2-Amino-3-bromo-5-methylbenzoic acid 98+%
2-Amino-3-bromo-5-methylbenzoic acid
Purity: 98+%
CAS No.: 13091-43-5
Molecular Formula: C8H8BrNO2
Formula Weight: 230.07
Melting Point: 202-206°
Beilstein Reference: 3540992
Formula:
C₈H₈BrNO₂ MW: 230.06 g/mol Melting Pt: 202…206 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051705 CAS Number: 13091-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...