1-Chloronaphthalene tech. 85% remainder 2-chloronaphthalene, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Triisodecyl phosphite, α-Chloronaphthalene

A15029-30 A15029-18 A15029-0B
AAAA15029-30EA 86.2 USD
AAAA15029-30 AAA-A15029-18 AAA-A15029-0B
1-Chloronaphthalene tech. 85% remainder 2-chloronaphthalene, Technical Grade
1-Chloronaphthalene

Purity: 85% CAS No.: 90-13-1 Molecular Formula: C10H7Cl Formula Weight: 162.62 Flash Point: 121°(249°F) Refractive Index: 1.6330 Melting Point: ca -8° Boiling Point: 259° Beilstein Reference: 970836 Merck Reference: 14,2149 MDL No.: MFCD00003874

Formula: C₁₀H₇Cl
Boiling Pt: 259 °C
Melting Pt: ∼ 8 °C
Density: 1.1938 g/cm³ (20 °C)
Flash Pt: 121 °C (249 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003874
CAS Number: 90-13-1
EINECS: 201-967-3
UN: 3082
ADR: 9,III
Merck Index: 13,02168

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR