Terephthalaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde
A14930-36
A14930-22
A14930-14
AAAA14930-36EA
228
USD
AAAA14930-36
AAAA14930-22
AAA14930-14
Terephthalaldehyde 98%
Terephthalaldehyde
Purity: 98%
CAS No.: 623-27-8
Molecular Formula: C8H6O2
Formula Weight: 134.14
Flash Point: 76°(168°F)
Melting Point: 114-117°
Boiling Point: 245°
Beilstein Reference: 385863
MDL No.: MFCD00006949
Formula:
C₈H₆O₂ MW: 134.13 g/mol Boiling Pt: 245 °C Melting Pt: 114…117 °C Density: 1.381 g/cm³ (20 °C) Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006949 CAS Number: 623-27-8 EINECS: 210-784-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...