2-(Trifluoromethyl)phenylhydrazine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-(Trifluoromethyl)phenyl)hydrazine, 2-Hydrazinobenzotrifluoride

A14781-06 A14781-03
AAA14781-06EA 158 USD
AAA14781-06 AAA14781-03
2-(Trifluoromethyl)phenylhydrazine 97%
2-(Trifluoromethyl)phenylhydrazine

Purity: 97% CAS No.: 365-34-4 Molecular Formula: C7H7F3N2 Formula Weight: 176.14 Melting Point: 57-63° Beilstein Reference: 641592

Formula: C₇H₇F₃N₂
MW: 176.14 g/mol
Melting Pt: 57…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00052687
CAS Number: 365-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR