2-Ethoxybenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A14758-22 A14758-36 A14758-14
AAAA14758-22EA 69.4 USD
AAAA14758-22 AAAA14758-36 AAAA14758-14
2-Ethoxybenzoic acid 98%
2-Ethoxybenzoic acid

Purity: 98% CAS No.: 134-11-2 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Refractive Index: 1.5410 Melting Point: 19-20° Boiling Point: 174-175°/15mm Beilstein Reference: 2691039 MDL No.: MFCD00002438

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 174…175 °C/15 mm
Melting Pt: 19…20 °C
Density: 1.173 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002438
CAS Number: 134-11-2
EINECS: 205-130-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR