4-Methylbiphenyl 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methyl-1,1'-biphenyl, 4-Phenyltoluene

A14702-03 A14702-06 A14702-14
AAAA14702-03EA 37.83 USD
AAAA14702-03 AAAA14702-06 AAAA14702-14
4-Methylbiphenyl 98%
4-Methylbiphenyl

Purity: 98% CAS No.: 644-08-6 Molecular Formula: C13H12 Formula Weight: 168.24 Flash Point: 110°(230°F) Melting Point: 44-48° Boiling Point: 267-268° Beilstein Reference: 1904468 MDL No.: MFCD00008544

Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 267…268 °C
Melting Pt: 44…48 °C
Density: 1.015 g/cm³ (27 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008544
CAS Number: 644-08-6
EINECS: 211-409-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR