3-Fluoroaniline 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14228-14
A14228-22
A14228-36
AAA14228-14EA
52.4
USD
AAA14228-14
AAA14228-22
AAA14228-36
3-Fluoroaniline 98+%
3-Fluoroaniline
Purity: 98+%
CAS No.: 372-19-0
Molecular Formula: C6H6FN
Formula Weight: 111.12
Flash Point: 77°(170°F)
Refractive Index: 1.5440
Odor: Recognizable
Boiling Point: 186-187°
Beilstein Reference: 1305471
UN No.: 2941
MDL No.: MFCD00007758
Formula:
C₆H₆FN MW: 111.12 g/mol Boiling Pt: 186…187 °C Melting Pt: -2 °C Density: 1.16 g/cm³ (20 °C) Flash Pt: 77 °C (170 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007758 CAS Number: 372-19-0 EINECS: 206-747-0 UN: 2941 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...