p-Tolylhydrazinium chloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methylphenylhydrazinium chloride, 4-Tolylhydrazine hydrochloride, p-Tolylhydrazine hydrochloride, 1-(4-Methylphenyl)hydrazine hydrochloride, 4-Methylphenylhydrazine hydrochloride, 1-(4-Tolyl)hydrazine hydrochloride
A14220-06
A14220-14
A14220-22
AAA14220-06EA
50.2
USD
AAA14220-06
AAA14220-14
AAA14220-22
p-Tolylhydrazinium chloride 98%
p-Tolylhydrazinium chloride
Purity: 98%
CAS No.: 637-60-5
Molecular Formula: C7H10N2.HCl
Formula Weight: 158.63
Melting Point: ca 240° dec.
Beilstein Reference: 3563996
UN No.: 2811
MDL No.: MFCD00012940
Formula:
C₇H₁₁ClN₂ Melting Pt: ca 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012940 CAS Number: 637-60-5 EINECS: 211-295-2 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...