3-Aminobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

A13995-09 A13995-18
AAAA13995-09EA 54.3 USD
AAAA13995-09 AAAA13995-18
3-Aminobenzonitrile 99%
3-Aminobenzonitrile

Purity: 99% CAS No.: 2237-30-1 Molecular Formula: C7H6N2 Formula Weight: 118.14 Flash Point: 110°(230°F) Odor: Almond-like Boiling Point: 290° UN No.: 3439 MDL No.: MFCD00007756

Formula: C₇H₆N₂
Boiling Pt: 290 °C
Melting Pt: 51…53 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007756
CAS Number: 2237-30-1
EINECS: 218-800-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR