1-Butylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Butylboronic acid, n-Butylboronic acid
A13725-14
A13725-03
A13725-06
AAA13725-14EA
366
USD
AAA13725-14
AAA13725-03
AAA13725-06
1-Butylboronic acid 98%
1-Butylboronic acid
Purity: 98%
CAS No.: 4426-47-5
Molecular Formula: CH3CH2CH2CH2B(OH)2
Formula Weight: 101.94
Storage Temperature: 2°C to 8°C
Melting Point: 92-96°
Beilstein Reference: 1733489
MDL No.: MFCD00002106
Formula:
C₄H₁₁BO₂ MW: 101.94 g/mol Melting Pt: 92…96 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002106 CAS Number: 4426-47-5 EINECS: 224-607-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...