Tetrahydropyran 98+%

Supplier: Thermo Scientific Chemicals
Danger

A13392-AP A13392-AE
AAA13392-APEA 206 USD
AAA13392-AP AAA13392-AE AAA13392-0F
Tetrahydropyran 98+%
Tetrahydropyran

Purity: 98+% CAS No.: 142-68-7 Molecular Formula: C5H10O Formula Weight: 86.14 Flash Point: -15°(5°F) Refractive Index: 1.4210 Melting Point: -49° Boiling Point: 87-89° Beilstein Reference: 102436 Merck Reference: 14,9217 UN No.: 3271 MDL No.: MFCD00006585

Formula: C₅H₁₀O
MW: 86.13 g/mol
Boiling Pt: 87…89 °C
Melting Pt: –49 °C
Density: 0.881 g/cm³ (20 °C)
Flash Pt: –15 °C (5 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006585
CAS Number: 142-68-7
EINECS: 205-552-8
UN: 3271
ADR: 3,III
Merck Index: 12,09356

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR