Tetramethylammonium bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

A13368-22 A13368-0E A13368-36
AAAA13368-22EA 39.2 USD
AAAA13368-22 AAAA13368-0E AAAA13368-36
Tetramethylammonium bromide 98%
Tetramethylammonium bromide

Purity: 98% CAS No.: 64-20-0 Molecular Formula: C4H12BrN Formula Weight: 154.06 Odor: Odorless Melting Point: 300° Beilstein Reference: 3620955 UN No.: 2811 MDL No.: MFCD00011626

Formula: C₄H₁₂BrN
MW: 154.05 g/mol
Boiling Pt: ∼360 °C (1013 hPa)
Melting Pt: >300 °C
Density: 1.56
Storage Temperature: Ambient
MDL Number: MFCD00011626
CAS Number: 64-20-0
EINECS: 200-581-2
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR