Tetramethylammonium chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TMACl, TMA

A13288-0B A13288-30 A13288-0I
AAAA13288-0BEA 150 USD
AAAA13288-0B AAAA13288-30 AAAA13288-0I
Tetramethylammonium chloride 97%
Tetramethylammonium chloride

Phase transfer catalyst


Purity: 97% CAS No.: 75-57-0 Molecular Formula: C4H12ClN Formula Weight: 109.60 Odor: Odorless Melting Point: 300° Beilstein Reference: 2496575 UN No.: 2811 MDL No.: MFCD00011628

Formula: (CH₃)₄NCl
Melting Pt: >300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR