2-(4-Fluorophenyl)indole 99%

Supplier: Thermo Scientific Chemicals
Warning

A13255-03 A13255-06
AAA13255-03EA 26.8 USD
AAA13255-03 AAA13255-06
2-(4-Fluorophenyl)indole 99%
2-(4-Fluorophenyl)indole

Purity: 99% CAS No.: 782-17-2 Molecular Formula: C14H10FN Formula Weight: 211.24 Odor: Odorless Melting Point: 186-191° Beilstein Reference: 744766 MDL No.: MFCD00068178

Formula: C₁₄H₁₀FN
Melting Pt: 186…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00068178
CAS Number: 782-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR