Naphthalene 99+%

Supplier: Thermo Scientific Chemicals
Warning

A13188-0B A13188-0I A13188-30
AAAA13188-0BEA 59.19 USD
AAAA13188-0B AAAA13188-0I AAAA13188-30
Naphthalene 99+%
Naphthalene

Purity: 99+% CAS No.: 91-20-3 Molecular Formula: C10H8 Formula Weight: 128.17 Flash Point: 78°(172°F) Refractive Index: 1.5821 Odor: Aromatic Melting Point: 80-82° Boiling Point: 218° Beilstein Reference: 1421310 Merck Reference: 14,6370 UN No.: 1334 MDL No.: MFCD00001742

Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218 °C
Melting Pt: 80…82 °C
Density: 1.145 g/cm³ (20 °C)
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1,III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR