Methyl acrylate 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ∼15 ppm
Purity: 99%
CAS No.: 96-33-3
Molecular Formula: CH2=CHCO2CH3
Formula Weight: 86.09
Storage Temperature: 2°C to 8°C
Flash Point: -3°(26°F)
Refractive Index: 1.4030
Odor: Stench
Melting Point: -75°
Boiling Point: 79-81°
Beilstein Reference: 605396
Merck Reference: 14,6011
UN No.: 1919
MDL No.: MFCD00008627
Formula:
C₄H₆O₂ Boiling Pt: 79…81 °C Melting Pt: –75 °C Density: 0.956 Flash Pt: -3 °C(26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008627 CAS Number: 96-33-3 EINECS: 202-500-6 UN: 1919 ADR: "3",II Merck Index: 14,6011 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...