Acetylferrocene 97%

Supplier: Thermo Scientific Chemicals
Danger

A13078-14 A13078-22
AAAA13078-14EA 303 USD
AAAA13078-14 AAAA13078-22
Acetylferrocene 97%
Acetylferrocene

Purity: 97% CAS No.: 1271-55-2 Molecular Formula: C12H12FeO Formula Weight: 228.07 Melting Point: 79-86° Boiling Point: 160-163°/4mm UN No.: 3467 MDL No.: MFCD00001432

Formula: C₁₂H₁₂FeO
Boiling Pt: 160…163 °C/4 mm
Melting Pt: 79…86 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001432
CAS Number: 1271-55-2
EINECS: 215-043-2
UN: 3467
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR