2,3-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Naphthalenediamine, DAN, 2,3-naphthylenediamine

A12993-06 A12993-14 A12993-03
AAA12993-06EA 459.97 USD
AAA12993-06 AAA12993-14 AAA12993-03
2,3-Diaminonaphthalene 97%
2,3-Diaminonaphthalene

MDL: MFCD00004116 Beilstein: 2206394 Slightly soluble in water


Purity: 97% CAS No.: 771-97-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 193-196° Solubility: Slightly soluble in water Beilstein Reference: 2206394 MDL No.: MFCD00004116

Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1.0968 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004116
CAS Number: 771-97-1
EINECS: 212-241-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR