Calcium hydroxide 95%

Supplier: Thermo Scientific Chemicals
Danger

A12650-0B A12650-0I A12650-30
AAA-A12650-0BEA 58.1 USD
AAA-A12650-0B AAA-A12650-0I AAA-A12650-30
Calcium hydroxide 95%
Calcium hydroxide
Formula: Ca(OH)₂
Melting Pt: 550 °C
Density: 2.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
ADR: 8,III
Merck Index: 13,01675

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR