1-Bromo-2-chloroethane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethylene chlorobromide
A12349-0E
A12349-36
A12349-22
AAA12349-0EEA
865
USD
AAA12349-0E
AAA12349-36
AAA12349-22
1-Bromo-2-chloroethane 98%
1-Bromo-2-chloroethane
Purity: 98%
CAS No.: 107-04-0
Molecular Formula: C2H4BrCl
Formula Weight: 143.42
Flash Point: None
Refractive Index: 1.4900
Odor: Sweet
Melting Point: -18°
Boiling Point: 106-107°
Beilstein Reference: 605265
UN No.: 2810
MDL No.: MFCD00000962
Formula:
BrH₂CCH₂Cl MW: 143.41 g/mol Boiling Pt: 106…107 °C Melting Pt: –18 °C Density: 1.7392 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000962 CAS Number: 107-04-0 EINECS: 203-456-0 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...