Benzophenone hydrazone 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12266-0B A12266-30 A12266-18
AAA12266-0BEA 272 USD
AAA12266-0B AAA12266-30 AAA12266-18
Benzophenone hydrazone 98+%
Benzophenone hydrazone

Purity: 98+% CAS No.: 5350-57-2 Molecular Formula: C13H12N2 Formula Weight: 196.25 Flash Point: 146°(294°F) Odor: Characteristic Boiling Point: 225-230°/55mm Beilstein Reference: 1910177 UN No.: 2811 MDL No.: MFCD00007624

Formula: C₁₃H₁₂N₂
MW: 196.25 g/mol
Boiling Pt: 225…230 °C/55 mm
Melting Pt: 96…100 °C
Density: 1.1
Flash Pt: 146 °C(294 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007624
CAS Number: 5350-57-2
EINECS: 226-321-8
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR