2,6-Dibromotoluene 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11832-14 A11832-03 A11832-06
AAA11832-14EA 506 USD
AAA11832-14 AAA11832-03 AAA11832-06
2,6-Dibromotoluene 98+%
2,6-Dibromotoluene

Purity: 98+% CAS No.: 69321-60-4 Molecular Formula: C7H6Br2 Formula Weight: 249.94 Refractive Index: 1.6060 Melting Point: 2-6° Boiling Point: 112-113°/7mm Beilstein Reference: 3235502 MDL No.: MFCD00013524

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Boiling Pt: 112…113 °C/7 mm
Melting Pt: 2…6 °C
Density: 1.812
Storage Temperature: Ambient
MDL Number: MFCD00013524
CAS Number: 69321-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR