L(+)-Lithium lactate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt

A11818-0B A11818-18 A11818-30
AAAA11818-0BEA 610 USD
AAAA11818-0B AAAA11818-18 AAAA11818-30
L(+)-Lithium lactate 97%
L(+)-Lithium lactate

Useful chiral synthon, building block for depsipeptides


Purity: 97% CAS No.: 27848-80-2 Molecular Formula: C3H5LiO3 Formula Weight: 96.01 Melting Point: 253-255° Beilstein Reference: 3568332

Formula: C₃H₅LiO₃
MW: 96.01 g/mol
Melting Pt: 253…255 °C
Storage Temperature: Ambient
MDL Number: MFCD00065512
CAS Number: 27848-80-2
EINECS: 248-692-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR