Methyl-6-quinolyl ether 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxyquinoline

A11736-06 A11736-14 A11736-22
AAA11736-06EA 30.6 USD
AAA11736-06 AAA11736-14 AAA11736-22
Methyl-6-quinolyl ether 98%
Methyl-6-quinolyl ether

Purity: 98% CAS No.: 5263-87-6 Molecular Formula: C10H9NO Formula Weight: 159.19 Flash Point: 110°(230°F) Refractive Index: 1.6250 Melting Point: 17-20° Boiling Point: 141-145°/15mm Beilstein Reference: 115244 MDL No.: MFCD00006800

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Boiling Pt: 141…145 °C/15 mm
Melting Pt: 17…20 °C
Density: 1.152 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006800
CAS Number: 5263-87-6
EINECS: 226-077-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR