Diboron trioxide 99%

Supplier: Thermo Scientific Chemicals
Danger

A11707-0E A11707-36
AAA11707-0EEA 408.98 USD
AAA11707-0E AAAA11707-36
Diboron trioxide 99%
Diboron trioxide

Purity: 99% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Odor: Odorless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315

Formula: B₂O₃
Boiling Pt: 1860 °C (1013 hPa)
Melting Pt: 450 °C
Density: 1.81 to 1.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR