Allyl trifluoroacetate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11690-18
A11690-09
AAA11690-18EA
140
USD
AAA11690-18
AAA11690-09
AAA11690-30
Allyl trifluoroacetate 97%
Allyl trifluoroacetate
Purity: 97%
CAS No.: 383-67-5
Molecular Formula: CH2=CHCH2O2CCF3
Formula Weight: 154.09
Flash Point: -1°(30°F)
Refractive Index: 1.3360
Boiling Point: 73-75°
Beilstein Reference: 1766312
UN No.: 2924
MDL No.: MFCD00013567
Formula:
C₅H₅F₃O₂ Boiling Pt: 73…75 °C Density: 1.183 Flash Pt: -1 °C(30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013567 CAS Number: 383-67-5 EINECS: 206-853-7 UN: 2924 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...