1,2-Bis(diphenylphosphino)ethane 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethylenebis(diphenylphosphine), Bis(1,2-diphenylphosphino)ethane, DIPHOS, 1,2-Bis(diphenylphosphino)ethane, Ethane-1,3-diylbis(diphenylphosphine)

A11419-06 A11419-14 A11419-22
AAAA11419-06EA 52.34 USD
AAAA11419-06 AAAA11419-14 AAAA11419-22
1,2-Bis(diphenylphosphino)ethane 97+%
1,2-Bis(diphenylphosphino)ethane

Purity: 97+% CAS No.: 1663-45-2 Molecular Formula: C26H24P2 Formula Weight: 398.42 Melting Point: 138-143° Beilstein Reference: 761261

Formula: C₂₆H₂₄P₂
Melting Pt: 138…143 °C
Storage Temperature: Ambient
MDL Number: MFCD00003047
CAS Number: 1663-45-2
EINECS: 216-769-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR