3,5-Dimethylphenylhydrazinium chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(3,5-Dimethylphenyl)hydrazine hydrochloride, 3,5-Dimethylphenylhydrazine hydrochloride

A11336-06 A11336-14
AAA11336-06EA 44.6 USD
AAA11336-06 AAA11336-14 AAA11336-22
3,5-Dimethylphenylhydrazinium chloride 98%
3,5-Dimethylphenylhydrazinium chloride

Purity: 98% CAS No.: 60481-36-9 Molecular Formula: C8H12N2.HCl Formula Weight: 172.66 Melting Point: 180° dec. MDL No.: MFCD00052269

Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
Melting Pt: 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00052269
CAS Number: 60481-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR