2-Acetylthiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(Thiazol-2-yl)-1-ethanone

A11266-06 A11266-03
AAAA11266-06EA 173 USD
AAAA11266-06 AAAA11266-03
2-Acetylthiazole 99%
2-Acetylthiazole

Purity: 99% CAS No.: 24295-03-2 Molecular Formula: C5H5NOS Formula Weight: 127.17 Flash Point: 78°(172°F) Refractive Index: 1.5480 Odor: Characteristic Boiling Point: 89-91°/12mm Beilstein Reference: 109803 MDL No.: MFCD00005324

Formula: C₅H₅NOS
MW: 127.17 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 65.5 °C
Density: 1.22 g/cm³ (20 °C)
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005324
CAS Number: 24295-03-2
EINECS: 246-134-5
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR