Phosphorous acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A11189-0B A11189-0I A11189-30
AAAA11189-0BEA 104 USD
AAAA11189-0B AAAA11189-0I AAAA11189-30
Phosphorous acid 98+%
Phosphorous acid

Air Sensitive & Hygroscopic


Purity: 98+% CAS No.: 13598-36-2 Molecular Formula: H3PO3 Formula Weight: 82.00 Odor: Odorless Melting Point: 74° Boiling Point: 200° dec. Merck Reference: 14,7346 UN No.: 2834 MDL No.: MFCD00137258

Formula: H₃O₃P
Boiling Pt: 200 °C (dec.)
Melting Pt: 74 °C
Density: 1.651
Storage Temperature: Ambient
MDL Number: MFCD00137258
CAS Number: 13598-36-2
EINECS: 237-066-7
UN: 2834
ADR: 8,III
Merck Index: 14,7346

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR