Molybdenum(VI) oxide 99.5%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Molybdenum trioxide

A11159-30 A11159-0B A11159-18
AAA11159-30EA 150.76 USD
AAA11159-30 AAA11159-0B AAA11159-18
Molybdenum(VI) oxide 99.5%
Molybdenum(VI) oxide

Purity: 99.5% CAS No.: 1313-27-5 Molecular Formula: MoO3 Formula Weight: 143.94 Odor: Odorless Melting Point: 795° Boiling Point: 1155° subl. Merck Reference: 14,6239 UN No.: 3288 MDL No.: MFCD00003469

Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: 1155 °C (1013 hPa)
Melting Pt: 795 °C
Density: 4.69 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
ADR: 6.1,III
Merck Index: 13,06261

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR