tert-Butylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Purity: 98%
CAS No.: 75-64-9
Molecular Formula: (CH3)3CNH2
Formula Weight: 73.14
Flash Point: -38°(-36°F)
Refractive Index: 1.3780
Odor: Amine-like
Melting Point: -67°
Boiling Point: 44-46°
Beilstein Reference: 605267
Merck Reference: 14,1545
UN No.: 2733
MDL No.: MFCD00008050
Formula:
C₄H₁₁N Boiling Pt: 44…46 °C Melting Pt: –67 °C Density: 0.6965 g/cm³ (20 °C) Flash Pt: –38 °C (–36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008050 CAS Number: 75-64-9 EINECS: 200-888-1 UN: 2734 ADR: 8,II Merck Index: 13,01544 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...