Benzenesulfonamide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10891-30 A10891-0B
AAAA10891-0IEA 523.89 USD
AAAA10891-0I AAAA10891-30 AAAA10891-0B
Benzenesulfonamide 98+%
Benzenesulphonamide

Purity: 98+% CAS No.: 98-10-2 Molecular Formula: C6H7NO2S Formula Weight: 157.19 Flash Point: 250°(482°F) Odor: Odorless Beilstein Reference: 1100566 MDL No.: MFCD00007930

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
Melting Pt: 151…154 °C
Flash Pt: 250 °C(482 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007930
CAS Number: 98-10-2
EINECS: 202-637-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR