Antimony(III) potassium oxitartrate hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A10889-22 A10889-0E A10889-36
AAAA10889-22EA 68.2 USD
AAAA10889-22 AAAA10889-0E AAAA10889-36
Antimony(III) potassium oxitartrate hydrate 98%
Antimony(III) potassium oxitartrate hydrate

Purity: 98% CAS No.: 331753-56-1 Molecular Formula: C8H4K2O12Sb2.xH2O Formula Weight: 613.82(anhy) Physical Form: Powder Density: 2.607 Appearance: White UN No.: 1551 MDL No.: MFCD00148863

Formula: C₈H₄K₂O₁₂Sb₂·H₂O
Density: 2.607
Storage Temperature: Ambient
MDL Number: MFCD00148863
CAS Number: 331753-56-1
EINECS: 608-190-2
UN: 1551
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR